Product Details

SNP ID
rs114340064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185164770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGTTAGTCTTAGGAAAGGTG[C/T]TGTAAGTTGAAATAAAATACACTAA
Phenotype
MIM: 616047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP97 PubMed Links

Gene Details

Gene
CFAP97
Gene Name
cilia and flagella associated protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292033.1 Intron NP_001278962.1
NM_020827.2 Intron NP_065878.1
XM_017008483.1 Intron XP_016863972.1
XM_017008484.1 Intron XP_016863973.1
XM_017008485.1 Intron XP_016863974.1
XM_017008486.1 Intron XP_016863975.1

View Full Product Details