Product Details

SNP ID
rs116240990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185397260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGGAAGCGGCCTTGCTTGCTT[G/T]CTTCTCTGGCAGCTGCAAACGGGCG
Phenotype
MIM: 611482
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD37 PubMed Links

Gene Details

Gene
ANKRD37
Gene Name
ankyrin repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181726.3 221 Missense Mutation TTG,TTT L46F NP_859077.1
XM_017008176.1 221 Missense Mutation TTG,TTT L46F XP_016863665.1
Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
There are no transcripts associated with this gene.

Gene
LRP2BP
Gene Name
LRP2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018409.3 221 Intron NP_060879.2
XM_005263125.4 221 Intron XP_005263182.1
XM_006714260.3 221 Intron XP_006714323.1
XM_006714265.3 221 Intron XP_006714328.1
XM_011532110.2 221 Intron XP_011530412.1
XM_011532111.2 221 Intron XP_011530413.1
XM_017008410.1 221 Intron XP_016863899.1
XM_017008411.1 221 Intron XP_016863900.1
Gene
UFSP2
Gene Name
UFM1 specific peptidase 2
There are no transcripts associated with this gene.

View Full Product Details