Product Details

SNP ID
rs111650413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87802726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCCTCATTTAAAACGATTTCCA[A/G]TTCAGGTAAATATAGAAATTCATTT
Phenotype
MIM: 147563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IBSP PubMed Links

Gene Details

Gene
IBSP
Gene Name
integrin binding sialoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004967.3 276 Missense Mutation ATT,GTT I60V NP_004958.2

View Full Product Details