Product Details
- SNP ID
-
rs116035805
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:88728564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACTTGGAATCAGTGTCTCTCTTG[C/T]TGATGAGCACCTCCAGGAGCCTCAG
- Phenotype
-
MIM: 613299
MIM: 613300
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM13A
PubMed Links
Gene Details
- Gene
- FAM13A
- Gene Name
- family with sequence similarity 13 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001015045.2 |
4961 |
Missense Mutation |
AAC,AGC |
N688S |
NP_001015045.1 |
NM_001265578.1 |
4961 |
Missense Mutation |
AAC,AGC |
N674S |
NP_001252507.1 |
NM_001265579.1 |
4961 |
Missense Mutation |
AAC,AGC |
N660S |
NP_001252508.1 |
NM_001265580.1 |
4961 |
Missense Mutation |
AAC,AGC |
N660S |
NP_001252509.1 |
NM_014883.3 |
4961 |
Missense Mutation |
AAC,AGC |
N1014S |
NP_055698.2 |
XM_005262681.2 |
4961 |
Missense Mutation |
AAC,AGC |
N1000S |
XP_005262738.1 |
XM_005262682.2 |
4961 |
Missense Mutation |
AAC,AGC |
N994S |
XP_005262739.1 |
XM_005262683.2 |
4961 |
Missense Mutation |
AAC,AGC |
N986S |
XP_005262740.1 |
XM_005262684.3 |
4961 |
Missense Mutation |
AAC,AGC |
N805S |
XP_005262741.1 |
XM_006714057.3 |
4961 |
Missense Mutation |
AAC,AGC |
N825S |
XP_006714120.1 |
XM_011531516.1 |
4961 |
Missense Mutation |
AAC,AGC |
N1014S |
XP_011529818.1 |
XM_011531517.1 |
4961 |
Missense Mutation |
AAC,AGC |
N986S |
XP_011529819.1 |
XM_011531518.1 |
4961 |
Missense Mutation |
AAC,AGC |
N828S |
XP_011529820.1 |
XM_011531519.2 |
4961 |
Missense Mutation |
AAC,AGC |
N828S |
XP_011529821.1 |
XM_017007624.1 |
4961 |
Missense Mutation |
AAC,AGC |
N972S |
XP_016863113.1 |
XM_017007625.1 |
4961 |
Missense Mutation |
AAC,AGC |
N959S |
XP_016863114.1 |
XM_017007626.1 |
4961 |
Missense Mutation |
AAC,AGC |
N837S |
XP_016863115.1 |
XM_017007627.1 |
4961 |
Missense Mutation |
AAC,AGC |
N805S |
XP_016863116.1 |
XM_017007628.1 |
4961 |
Missense Mutation |
AAC,AGC |
N763S |
XP_016863117.1 |
XM_017007629.1 |
4961 |
Missense Mutation |
AAC,AGC |
N742S |
XP_016863118.1 |
XM_017007630.1 |
4961 |
Missense Mutation |
AAC,AGC |
N742S |
XP_016863119.1 |
XM_017007631.1 |
4961 |
Missense Mutation |
AAC,AGC |
N728S |
XP_016863120.1 |
XM_017007632.1 |
4961 |
Missense Mutation |
AAC,AGC |
N728S |
XP_016863121.1 |
XM_017007633.1 |
4961 |
Missense Mutation |
AAC,AGC |
N693S |
XP_016863122.1 |
XM_017007634.1 |
4961 |
Missense Mutation |
AAC,AGC |
N646S |
XP_016863123.1 |
- Gene
- FAM13A-AS1
- Gene Name
- FAM13A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details