Product Details

SNP ID
rs116168464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105716819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAATGAAGAAAGTTAATGAGAC[A/G]AACTCTGAGACATAAAGGAGGATTA
Phenotype
MIM: 615912 MIM: 611355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTCD PubMed Links

Gene Details

Gene
GSTCD
Gene Name
glutathione S-transferase C-terminal domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031720.3 Intron NP_001026890.2
NM_024751.3 Intron NP_079027.2
XM_005263222.3 Intron XP_005263279.1
XM_011532248.2 Intron XP_011530550.1
XM_011532249.2 Intron XP_011530551.1
XM_011532252.2 Intron XP_011530554.1
Gene
INTS12
Gene Name
integrator complex subunit 12
There are no transcripts associated with this gene.

View Full Product Details