Product Details

SNP ID
rs118168968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107992646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGACTTTACCTCTGTGTCTC[A/G]GTTTTCTCATCTCTAAAGTAGACTA
Phenotype
MIM: 601609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HADH PubMed Links

Gene Details

Gene
HADH
Gene Name
hydroxyacyl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184705.2 Intron NP_001171634.2
NM_005327.4 Intron NP_005318.3
XM_005262972.2 Intron XP_005263029.1

View Full Product Details