Product Details

SNP ID
rs113479481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118279388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAACCGTTGTTCAATGTTTCTA[A/G]GAAGCTGAGCTGAACACATTACGAT
Phenotype
MIM: 606709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS12 PubMed Links

Gene Details

Gene
PRSS12
Gene Name
protease, serine 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003619.3 Intron NP_003610.2
XM_005263318.4 Intron XP_005263375.1
XM_011532387.2 Intron XP_011530689.1
Gene
SNHG8
Gene Name
small nucleolar RNA host gene 8
There are no transcripts associated with this gene.

Gene
SNORA24
Gene Name
small nucleolar RNA, H/ACA box 24
There are no transcripts associated with this gene.

View Full Product Details