Product Details

SNP ID
rs115252011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119318082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTCAGATGATCCTGATTTTCA[C/G]CCAAGCGTGGGAATGACTGATGTGT
Phenotype
MIM: 134640
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FABP2 PubMed Links

Gene Details

Gene
FABP2
Gene Name
fatty acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000134.3 1417 UTR 3 NP_000125.2

View Full Product Details