Product Details

SNP ID
rs114411398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119318774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTTAAAAGGCAAGGCTACAT[A/G]TAAAGCAACATGGACGCAATATTTA
Phenotype
MIM: 134640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FABP2 PubMed Links

Gene Details

Gene
FABP2
Gene Name
fatty acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000134.3 725 UTR 3 NP_000125.2

View Full Product Details