Product Details

SNP ID
rs116405363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:81030946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGGTGTTCCCAAAAATAAAGC[A/G]AGGAGGGAAGGTACAGACAGATCTT
Phenotype
MIM: 112263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP3 PubMed Links

Gene Details

Gene
BMP3
Gene Name
bone morphogenetic protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006714291.3 169 UTR 5 XP_006714354.1

View Full Product Details