Product Details

SNP ID
rs116010000
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:81092815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAGCTCTTGTAGTTGTTCTTT[A/G]CCTTATATCCTATCTGAGTCCATGA
Phenotype
MIM: 601591
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PRKG2 PubMed Links

Gene Details

Gene
PRKG2
Gene Name
protein kinase, cGMP-dependent, type II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282480.1 Intron NP_001269409.1
NM_001282481.1 Intron NP_001269410.1
NM_001282482.1 Intron NP_001269411.1
NM_001282483.1 Intron NP_001269412.1
NM_001282485.1 Intron NP_001269414.1
NM_006259.2 Intron NP_006250.1
XM_005263126.3 Intron XP_005263183.1
XM_017008413.1 Intron XP_016863902.1
XM_017008414.1 Intron XP_016863903.1
XM_017008415.1 Intron XP_016863904.1
XM_017008416.1 Intron XP_016863905.1
XM_017008417.1 Intron XP_016863906.1
XM_017008418.1 Intron XP_016863907.1

View Full Product Details