Product Details
- SNP ID
-
rs113106660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:140622661 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCGGGGGCCAGGCTGGCCGGCAGG[G/T]GCTCAACAGACTCCACCACGTAAGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D9
PubMed Links
Gene Details
- Gene
- TBC1D9
- Gene Name
- TBC1 domain family member 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015130.2 |
3607 |
Missense Mutation |
CAC,CCC |
H1112P |
NP_055945.2 |
View Full Product Details