Product Details

SNP ID
rs115728727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144111207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTTATCTTGAAATATTAATGA[A/G]CAGAGATTTTTTTCCCCATCATTGA
Phenotype
MIM: 111300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GYPA PubMed Links

Gene Details

Gene
GYPA
Gene Name
glycophorin A (MNS blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308187.1 681 UTR 3 NP_001295116.1
NM_001308190.1 681 UTR 3 NP_001295119.1
NM_002099.7 681 UTR 3 NP_002090.4
XM_017008134.1 681 UTR 3 XP_016863623.1
XM_017008135.1 681 UTR 3 XP_016863624.1
XM_017008136.1 681 UTR 3 XP_016863625.1

View Full Product Details