Product Details
- SNP ID
-
rs117902141
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:145680420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTCTTTTAGAGCAGGACAGCAT[G/T]AGCCTGAGTGTAAACAGTAAGATTT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C4orf51
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7683750] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4orf51
- Gene Name
- chromosome 4 open reading frame 51
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080531.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
NP_001074000.1 |
XM_006714289.3 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_006714352.1 |
XM_011532196.2 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_011530498.1 |
XM_011532197.2 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_011530499.1 |
XM_017008547.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864036.1 |
XM_017008548.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864037.1 |
XM_017008549.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864038.1 |
XM_017008550.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864039.1 |
XM_017008551.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864040.1 |
XM_017008552.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864041.1 |
XM_017008553.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864042.1 |
XM_017008554.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864043.1 |
XM_017008555.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864044.1 |
XM_017008556.1 |
318 |
Nonsense Mutation |
GAG,TAG |
E73* |
XP_016864045.1 |
View Full Product Details