Product Details

SNP ID
rs114932002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82353688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGACACATTATGGCAAAAATTA[A/G]ATTTTGCTAAAAACATTTCAAACTT
Phenotype
MIM: 601324
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPD PubMed Links

Gene Details

Gene
HNRNPD
Gene Name
heterogeneous nuclear ribonucleoprotein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003810.1 1736 UTR 3 NP_001003810.1
NM_002138.3 1736 UTR 3 NP_002129.2
NM_031369.2 1736 UTR 3 NP_112737.1
NM_031370.2 1736 UTR 3 NP_112738.1

View Full Product Details