Product Details

SNP ID
rs111456531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152769526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGCTAAAGAATTTTGAAGTCC[A/G]TCATTCATATCTTGACTTCTGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIGD4 PubMed Links

Gene Details

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 2300 Silent Mutation GAC,GAT D493D NP_663772.1
XM_005262807.4 2300 Silent Mutation GAC,GAT D493D XP_005262864.1

View Full Product Details