Product Details

SNP ID
rs113300275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152769827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAAATCCAGACCAGTATCCTTC[C/T]CTGCATTTGTTATGTCACTTTCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIGD4 PubMed Links

Gene Details

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 1999 Missense Mutation GAG,GGG E393G NP_663772.1
XM_005262807.4 1999 Missense Mutation GAG,GGG E393G XP_005262864.1

View Full Product Details