Product Details

SNP ID
rs116170607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153346353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACTGAGAAGGTTCCTCAGATAA[C/T]ATGATTTTTTAAGTGGTTTAAAATA
Phenotype
MIM: 611422 MIM: 614141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MND1 PubMed Links

Gene Details

Gene
MND1
Gene Name
meiotic nuclear divisions 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253861.1 Intron NP_001240790.1
NM_032117.3 Intron NP_115493.1
XM_005263275.2 Intron XP_005263332.1
XM_017008688.1 Intron XP_016864177.1
Gene
TRIM2
Gene Name
tripartite motif containing 2
There are no transcripts associated with this gene.

View Full Product Details