Product Details

SNP ID
rs112892166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82926374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATCTACATTTAAAGTTACTGC[A/G]CTATGATTTTATAAATCCAAATTTA
Phenotype
MIM: 613367 MIM: 612537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIN54 PubMed Links

Gene Details

Gene
LIN54
Gene Name
lin-54 DREAM MuvB core complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115007.2 3903 UTR 3 NP_001108479.1
NM_001115008.2 3903 UTR 3 NP_001108480.1
NM_001288996.1 3903 UTR 3 NP_001275925.1
NM_001288997.1 3903 UTR 3 NP_001275926.1
NM_194282.3 3903 UTR 3 NP_919258.2
XM_005262750.4 3903 UTR 3 XP_005262807.1
XM_006714081.3 3903 UTR 3 XP_006714144.1
XM_017007728.1 3903 UTR 3 XP_016863217.1
XM_017007729.1 3903 UTR 3 XP_016863218.1
XM_017007730.1 3903 UTR 3 XP_016863219.1
Gene
THAP9
Gene Name
THAP domain containing 9
There are no transcripts associated with this gene.

View Full Product Details