Product Details

SNP ID
rs111918853
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:161385907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTCCTGGATTTGCCTGTTTTTC[C/T]GGTTCCAAGGCCATTCTTCTGCCTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FSTL5 PubMed Links
Additional Information
For this assay, SNP(s) [rs17040984] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FSTL5
Gene Name
follistatin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128427.2 2817 Missense Mutation CAG,CGG Q794R NP_001121899.1
NM_001128428.2 2817 Missense Mutation CAG,CGG Q785R NP_001121900.1
NM_020116.4 2817 Missense Mutation CAG,CGG Q795R NP_064501.2
XM_011532126.1 2817 Missense Mutation CAG,CGG Q786R XP_011530428.1

View Full Product Details