Product Details

SNP ID
rs114876653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169098152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAATACATGCTTATTATTGTT[C/T]TAAGCCACTGACTTTTGGGCGTAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3RF1 PubMed Links

Gene Details

Gene
SH3RF1
Gene Name
SH3 domain containing ring finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020870.3 Intron NP_065921.2

View Full Product Details