Product Details

SNP ID
rs121434481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174493236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGTTTTCTTCTTTTTCAATTG[G/A]TTCAAGGATGGCTGTGTTAACAAAG
Phenotype
MIM: 601688
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
HPGD PubMed Links

Gene Details

Gene
HPGD
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000860.5 754 Missense Mutation CCA,TCA P193S NP_000851.2
NM_001145816.2 754 Intron NP_001139288.1
NM_001256301.1 754 Missense Mutation CCA,TCA P72S NP_001243230.1
NM_001256305.1 754 Intron NP_001243234.1
NM_001256306.1 754 Missense Mutation CCA,TCA P125S NP_001243235.1
NM_001256307.1 754 Missense Mutation CCA,TCA P72S NP_001243236.1
XM_011531907.1 754 Intron XP_011530209.1

View Full Product Details