Product Details

SNP ID
rs116768611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135937515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACGAGCCCTGGGTTCCCAGTGCC[C/T]TCCATCAAGAAAATACTCCACAACC
Phenotype
MIM: 608129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBAC1 PubMed Links

Gene Details

Gene
UBAC1
Gene Name
UBA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016172.2 Intron NP_057256.2

View Full Product Details