Product Details

SNP ID
rs111591069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119556546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCTAGTATTTTCTTATTGCTA[A/T]ATTTTTTTTCTTTTTTTTTTTTTTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP221 PubMed Links

Gene Details

Gene
CFAP221
Gene Name
cilia and flagella associated protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271049.1 Intron NP_001257978.1
XM_006712353.3 Intron XP_006712416.1
XM_017003550.1 Intron XP_016859039.1
XM_017003551.1 Intron XP_016859040.1
XM_017003552.1 Intron XP_016859041.1
XM_017003553.1 Intron XP_016859042.1
XM_017003554.1 Intron XP_016859043.1
XM_017003555.1 Intron XP_016859044.1
XM_017003556.1 Intron XP_016859045.1
XM_017003557.1 Intron XP_016859046.1
XM_017003558.1 Intron XP_016859047.1
XM_017003559.1 Intron XP_016859048.1

View Full Product Details