Product Details

SNP ID
rs115845526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119559708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGCATCTGGTCAATGTTTCCA[A/G]TGAAGACACACGTGTTCATATTTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP221 PubMed Links

Gene Details

Gene
CFAP221
Gene Name
cilia and flagella associated protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271049.1 302 Missense Mutation AAT,AGT N87S NP_001257978.1
XM_006712353.3 302 Missense Mutation AAT,AGT N87S XP_006712416.1
XM_017003550.1 302 Missense Mutation AAT,AGT N87S XP_016859039.1
XM_017003551.1 302 Missense Mutation AAT,AGT N87S XP_016859040.1
XM_017003552.1 302 Missense Mutation AAT,AGT N87S XP_016859041.1
XM_017003553.1 302 Missense Mutation AAT,AGT N87S XP_016859042.1
XM_017003554.1 302 UTR 5 XP_016859043.1
XM_017003555.1 302 UTR 5 XP_016859044.1
XM_017003556.1 302 UTR 5 XP_016859045.1
XM_017003557.1 302 UTR 5 XP_016859046.1
XM_017003558.1 302 Missense Mutation AAT,AGT N87S XP_016859047.1
XM_017003559.1 302 UTR 5 XP_016859048.1

View Full Product Details