Product Details

SNP ID
rs116648807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:137964493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCTTTCTCAGAAAACCAAATA[C/T]GGCATCTTCCATGAGGAGCTTGTTT
Phenotype
MIM: 605238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNMT PubMed Links

Gene Details

Gene
HNMT
Gene Name
histamine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024074.2 89 Missense Mutation ACG,ATG T1M NP_001019245.1
NM_001024075.1 89 Missense Mutation ACG,ATG T1M NP_001019246.1
NM_006895.2 89 Missense Mutation ACG,ATG T1M NP_008826.1
XM_011511064.2 89 UTR 5 XP_011509366.1
XM_017003948.1 89 UTR 5 XP_016859437.1
XM_017003949.1 89 Missense Mutation ACG,ATG T1M XP_016859438.1

View Full Product Details