Product Details

SNP ID
rs115518761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:146587339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCAGCAGACAGCGGCGGACAA[A/G]CGACCTCGCTCTGTGGCCAGTGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PABPC1P2 PubMed Links

Gene Details

Gene
PABPC1P2
Gene Name
poly(A) binding protein cytoplasmic 1 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details