Product Details

SNP ID
rs114098076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113709720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGCTGGGGCATTACCTGGAC[A/G]CTTGTTAGAAAGGCAAATTACCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35F5 PubMed Links

Gene Details

Gene
SLC35F5
Gene Name
solute carrier family 35 member F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263799.3 7537 UTR 3 XP_005263856.1
XM_011511921.2 7537 UTR 3 XP_011510223.1
XM_011511922.2 7537 UTR 3 XP_011510224.1
XM_011511923.2 7537 UTR 3 XP_011510225.1
XM_017005027.1 7537 Intron XP_016860516.1
XM_017005028.1 7537 UTR 3 XP_016860517.1

View Full Product Details