Product Details

SNP ID
rs118172849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96252303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAACTATCTATCACCTTAAAGGT[C/T]GGAAGAGATTGGAATCCATTTCCTT
Phenotype
MIM: 613403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STARD7-AS1 PubMed Links

Gene Details

Gene
STARD7-AS1
Gene Name
STARD7 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMEM127
Gene Name
transmembrane protein 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193304.2 2124 UTR 3 NP_001180233.1
NM_017849.3 2124 UTR 3 NP_060319.1
XM_017004450.1 2124 UTR 3 XP_016859939.1
XM_017004451.1 2124 UTR 3 XP_016859940.1
XM_017004452.1 2124 UTR 3 XP_016859941.1

View Full Product Details