Product Details

SNP ID
rs116519747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99185728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGACCACAAACTTTATTGATA[A/G]CTGGATGCACTTGGATAAGTTATTG
Phenotype
MIM: 611838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MITD1 PubMed Links

Gene Details

Gene
MITD1
Gene Name
microtubule interacting and trafficking domain containing 1
There are no transcripts associated with this gene.

Gene
MRPL30
Gene Name
mitochondrial ribosomal protein L30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145212.3 Intron NP_660213.1

View Full Product Details