Product Details
- SNP ID
-
rs116764744
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100471486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCGATTCTATTTAACTACTTCAA[C/T]GACCCCAACTCACTGCCAGCTATGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NMS
PubMed Links
Gene Details
- Gene
- NMS
- Gene Name
- neuromedin S
View Full Product Details