Product Details

SNP ID
rs116039205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:33727007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGCGGGGGCAGGATGAAGAGG[A/T]TGGAGAGGCAAACGGGGCCCTTTGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01317 PubMed Links

Gene Details

Gene
LINC01317
Gene Name
long intergenic non-protein coding RNA 1317
There are no transcripts associated with this gene.

Gene
MYADML
Gene Name
myeloid associated differentiation marker like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details