Product Details

SNP ID
rs114671056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:44172333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAGAATTGATGGTTAAGTGTAC[C/T]GACATTTCACTATGCTATACTCAAA
Phenotype
MIM: 603770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPM1B PubMed Links

Gene Details

Gene
PPM1B
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033557.2 Intron NP_001028729.1
NM_002706.5 Intron NP_002697.1
NM_177968.3 Intron NP_808907.1
NM_177969.3 Intron NP_808908.1
XM_011532936.2 Intron XP_011531238.1
XM_017004395.1 Intron XP_016859884.1
XM_017004396.1 Intron XP_016859885.1
XM_017004397.1 Intron XP_016859886.1

View Full Product Details