Product Details

SNP ID
rs116161895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46701318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCACTGGCTGTGAAGTAGTAGT[A/G]ATAGCTAAACAGAATGAGAATGGGA
Phenotype
MIM: 607094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOCS5 PubMed Links

Gene Details

Gene
SOCS5
Gene Name
suppressor of cytokine signaling 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014011.4 Intron NP_054730.1
NM_144949.2 Intron NP_659198.1

View Full Product Details