Product Details

SNP ID
rs115363888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:23693378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTTCGCGCTGCAGATCTTCCC[C/T]GAGGTGGCCGCCCAGGAGGAGATCC
Phenotype
MIM: 615347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD2B PubMed Links

Gene Details

Gene
ATAD2B
Gene Name
ATPase family, AAA domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242338.2 1799 Intron NP_001229267.2
NM_017552.3 1799 Intron NP_060022.2
XM_005264372.3 1799 Intron XP_005264429.2
XM_006712030.3 1799 Intron XP_006712093.1
XM_006712031.3 1799 Intron XP_006712094.1
XM_011532918.2 1799 Intron XP_011531220.1
XM_011532919.2 1799 Intron XP_011531221.1
XM_011532920.2 1799 Intron XP_011531222.1
XM_011532921.2 1799 Intron XP_011531223.1
XM_011532924.2 1799 Intron XP_011531226.1
XM_011532925.2 1799 Intron XP_011531227.1
XM_011532928.1 1799 Intron XP_011531230.1
XM_011532929.2 1799 Intron XP_011531231.1
XM_011532930.2 1799 Intron XP_011531232.1
XM_011532931.2 1799 Intron XP_011531233.1
XM_011532932.1 1799 Intron XP_011531234.1
XM_017004375.1 1799 Intron XP_016859864.1
Gene
KLHL29
Gene Name
kelch like family member 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052920.1 1799 Silent Mutation CCC,CCT P464P NP_443152.1
XM_006711929.3 1799 Silent Mutation CCC,CCT P464P XP_006711992.1
XM_011532501.2 1799 Silent Mutation CCC,CCT P158P XP_011530803.1
XM_017003264.1 1799 Silent Mutation CCC,CCT P464P XP_016858753.1

View Full Product Details