Product Details

SNP ID
rs114157453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:56186401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCACCAATACAATGCAATGATA[A/G]TAACTTTCTGAGGGTGTTATGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85A PubMed Links

Gene Details

Gene
CCDC85A
Gene Name
coiled-coil domain containing 85A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080433.1 Intron NP_001073902.1
XM_005264125.1 Intron XP_005264182.1
Gene
LOC100129434
Gene Name
uncharacterized LOC100129434
There are no transcripts associated with this gene.

Gene
LOC105374690
Gene Name
uncharacterized LOC105374690
There are no transcripts associated with this gene.

Gene
LOC107985795
Gene Name
uncharacterized LOC107985795
There are no transcripts associated with this gene.

View Full Product Details