Product Details

SNP ID
rs113130006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:67399223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGGACTCACTGTCATCTTCCT[G/T]CAGTTCTCCTAATGATCCAGATGGA
Phenotype
MIM: 613196
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ETAA1 PubMed Links

Gene Details

Gene
ETAA1
Gene Name
Ewing tumor associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019002.3 410 Missense Mutation TGC,TTC C93F NP_061875.2
XM_017004376.1 410 Missense Mutation TGC,TTC C93F XP_016859865.1
XM_017004377.1 410 Missense Mutation TGC,TTC C93F XP_016859866.1

View Full Product Details