Product Details
- SNP ID
-
rs115137514
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68159672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCATCATCCGGACTGGTATAGTCT[C/T]ATCTCTGTTAATGGAATATAATAAT
- Phenotype
-
MIM: 610729
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PNO1
PubMed Links
Gene Details
- Gene
- PNO1
- Gene Name
- partner of NOB1 homolog
There are no transcripts associated with this gene.
- Gene
- WDR92
- Gene Name
- WD repeat domain 92
There are no transcripts associated with this gene.
View Full Product Details