Product Details

SNP ID
rs115177776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69459600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGGTGAGATTGTCACAGACTT[C/G]GCTGAAACTTAACAAGAAACTGTTT
Phenotype
MIM: 616405
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AAK1 PubMed Links

Gene Details

Gene
AAK1
Gene Name
AP2 associated kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014911.3 19660 UTR 3 NP_055726.3

View Full Product Details