Product Details

SNP ID
rs117102458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70449110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAGGGGGTCAGCTCTGAACAGAT[C/T]CAAGTGCTTCAGGTGTGGCATTGTT
Phenotype
MIM: 190170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGFA PubMed Links

Gene Details

Gene
TGFA
Gene Name
transforming growth factor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099691.2 2644 UTR 3 NP_001093161.1
NM_001308158.1 2644 UTR 3 NP_001295087.1
NM_001308159.1 2644 UTR 3 NP_001295088.1
NM_003236.3 2644 UTR 3 NP_003227.1

View Full Product Details