Product Details

SNP ID
rs116185049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70986447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATGCAATGCATTTTAGGAAGA[C/T]AGCTCTGGCCACATGATAGAGAAGC
Phenotype
MIM: 617009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD53 PubMed Links

Gene Details

Gene
ANKRD53
Gene Name
ankyrin repeat domain 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115116.1 2858 Intron NP_001108588.1
NM_024933.3 2858 Intron NP_079209.3
XM_005264569.4 2858 Intron XP_005264626.1
XM_011533114.2 2858 Intron XP_011531416.1
Gene
TEX261
Gene Name
testis expressed 261
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144582.2 2858 UTR 3 NP_653183.2

View Full Product Details