Product Details

SNP ID
rs114990701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79518057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATCTGTTCAATTAGGGGTTTTC[C/T]AAATTTTTGCTGCGATGTCTACTAT
Phenotype
MIM: 114025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNA2 PubMed Links

Gene Details

Gene
CTNNA2
Gene Name
catenin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164883.1 Intron NP_001158355.1
NM_001282597.2 Intron NP_001269526.1
NM_001282598.1 Intron NP_001269527.1
NM_001282599.1 Intron NP_001269528.1
NM_001282600.1 Intron NP_001269529.1
NM_001320810.1 Intron NP_001307739.1
NM_004389.3 Intron NP_004380.2
XM_011532555.2 Intron XP_011530857.1
XM_011532556.2 Intron XP_011530858.1
XM_011532557.2 Intron XP_011530859.1
XM_017003403.1 Intron XP_016858892.1
XM_017003404.1 Intron XP_016858893.1
XM_017003405.1 Intron XP_016858894.1
XM_017003406.1 Intron XP_016858895.1

View Full Product Details