Product Details

SNP ID
rs111463426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88126196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCTTGACTTTCTCCCCTGTCA[C/T]TGTCTCCAGCTCACATTCCTCCCCC
Phenotype
MIM: 134650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP1 PubMed Links

Gene Details

Gene
FABP1
Gene Name
fatty acid binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001443.2 334 Missense Mutation ATG,GTG M74V NP_001434.1

View Full Product Details