Product Details

SNP ID
rs114563957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88692002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGAGGGTGAGCACTTCGAAACGT[A/G]GGGCGCGGGGCGCATGTCCTTGGCG
Phenotype
MIM: 180430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPIA PubMed Links

Gene Details

Gene
RPIA
Gene Name
ribose 5-phosphate isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144563.2 Intron NP_653164.2

View Full Product Details