Product Details

SNP ID
rs113352216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:1632320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTAAGAGGCACAGAACACTCC[C/G]AGAACCCAGAATCTGCTGTCATCTG
Phenotype
MIM: 605158
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PXDN PubMed Links

Gene Details

Gene
PXDN
Gene Name
peroxidasin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012293.2 6416 UTR 3 NP_036425.1
XM_005264707.3 6416 UTR 3 XP_005264764.1
XM_011510396.1 6416 UTR 3 XP_011508698.1
XM_011510397.2 6416 UTR 3 XP_011508699.1

View Full Product Details