Product Details

SNP ID
rs115095308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69111011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTAAAGGGTTTCAAGTGTCA[C/T]TGTATCTTAGGAAAGTAACTCATCA
Phenotype
MIM: 605709 MIM: 616112
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP5 PubMed Links

Gene Details

Gene
ARL6IP5
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
There are no transcripts associated with this gene.

Gene
LMOD3
Gene Name
leiomodin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304418.1 Intron NP_001291347.1
NM_198271.4 Intron NP_938012.2

View Full Product Details