Product Details
- SNP ID
-
rs113891864
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:70959385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGGGGCTCTTCTTTGACGTGTAC[A/T]GGATGCCTGGAAAAAATATGCAGAG
- Phenotype
-
MIM: 605515
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXP1
PubMed Links
Gene Details
- Gene
- FOXP1
- Gene Name
- forkhead box P1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012505.1 |
2360 |
Intron |
|
|
NP_001012523.1 |
NM_001244808.1 |
2360 |
Silent Mutation |
CCA,CCT |
P631P |
NP_001231737.1 |
NM_001244810.1 |
2360 |
Silent Mutation |
CCA,CCT |
P648P |
NP_001231739.1 |
NM_001244812.1 |
2360 |
Silent Mutation |
CCA,CCT |
P556P |
NP_001231741.1 |
NM_001244813.1 |
2360 |
Silent Mutation |
CCA,CCT |
P532P |
NP_001231742.1 |
NM_001244814.1 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
NP_001231743.1 |
NM_001244815.1 |
2360 |
Silent Mutation |
CCA,CCT |
P634P |
NP_001231744.1 |
NM_001244816.1 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
NP_001231745.1 |
NM_032682.5 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
NP_116071.2 |
XM_005264735.3 |
2360 |
Silent Mutation |
CCA,CCT |
P633P |
XP_005264792.1 |
XM_005264736.3 |
2360 |
Silent Mutation |
CCA,CCT |
P633P |
XP_005264793.1 |
XM_005264737.4 |
2360 |
Silent Mutation |
CCA,CCT |
P631P |
XP_005264794.1 |
XM_005264742.3 |
2360 |
Silent Mutation |
CCA,CCT |
P531P |
XP_005264799.1 |
XM_006713102.2 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_006713165.1 |
XM_006713103.2 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_006713166.1 |
XM_006713104.2 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_006713167.1 |
XM_011533584.2 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_011531886.1 |
XM_011533585.2 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_011531887.1 |
XM_011533588.2 |
2360 |
Silent Mutation |
CCA,CCT |
P532P |
XP_011531890.1 |
XM_017006165.1 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_016861654.1 |
XM_017006166.1 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_016861655.1 |
XM_017006167.1 |
2360 |
Silent Mutation |
CCA,CCT |
P632P |
XP_016861656.1 |
XM_017006168.1 |
2360 |
Intron |
|
|
XP_016861657.1 |
View Full Product Details