Product Details

SNP ID
rs114026554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15044093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATGCCCCCTTGAAAAGGGCTC[A/G]TGTTTCTGCAGCTCCATCATAACTG
Phenotype
MIM: 611987 MIM: 601426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS25 PubMed Links

Gene Details

Gene
MRPS25
Gene Name
mitochondrial ribosomal protein S25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022497.4 3238 Intron NP_071942.1
Gene
NR2C2
Gene Name
nuclear receptor subfamily 2 group C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291694.1 3238 UTR 3 NP_001278623.1
NM_003298.4 3238 UTR 3 NP_003289.2
XM_011534058.2 3238 UTR 3 XP_011532360.1
XM_011534059.2 3238 UTR 3 XP_011532361.1
XM_011534061.2 3238 UTR 3 XP_011532363.1
XM_011534063.2 3238 UTR 3 XP_011532365.1
XM_011534064.2 3238 UTR 3 XP_011532366.1
XM_011534065.2 3238 UTR 3 XP_011532367.1
XM_011534066.2 3238 UTR 3 XP_011532368.1
XM_017007118.1 3238 UTR 3 XP_016862607.1
XM_017007119.1 3238 UTR 3 XP_016862608.1
XM_017007120.1 3238 UTR 3 XP_016862609.1

View Full Product Details