Product Details

SNP ID
rs115192716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:21421058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCCCTCCCACCCCCAAACCT[C/T]CCCCACTTTTTTATAACCTTTTCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF385D PubMed Links

Gene Details

Gene
ZNF385D
Gene Name
zinc finger protein 385D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024697.2 1956 UTR 3 NP_078973.1
XM_011534122.1 1956 Intron XP_011532424.1
XM_011534123.2 1956 Intron XP_011532425.1
XM_011534124.2 1956 Intron XP_011532426.1
XM_017007191.1 1956 UTR 3 XP_016862680.1
XM_017007192.1 1956 UTR 3 XP_016862681.1
XM_017007193.1 1956 UTR 3 XP_016862682.1
XM_017007194.1 1956 UTR 3 XP_016862683.1
XM_017007195.1 1956 UTR 3 XP_016862684.1
XM_017007196.1 1956 UTR 3 XP_016862685.1
XM_017007197.1 1956 UTR 3 XP_016862686.1
XM_017007198.1 1956 UTR 3 XP_016862687.1
XM_017007199.1 1956 UTR 3 XP_016862688.1
XM_017007200.1 1956 UTR 3 XP_016862689.1
XM_017007201.1 1956 UTR 3 XP_016862690.1
XM_017007202.1 1956 UTR 3 XP_016862691.1
XM_017007203.1 1956 Intron XP_016862692.1

View Full Product Details