Product Details

SNP ID
rs111410709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23893092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATACAATTGTGGAAATTACTGA[C/T]TTTTAGTTCTCAGAATTAGAGTTCC
Phenotype
MIM: 604496 MIM: 602916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020345.3 823 UTR 3 NP_065078.1
XM_005265076.3 823 UTR 3 XP_005265133.1
XM_005265077.4 823 UTR 3 XP_005265134.1
XM_005265078.3 823 UTR 3 XP_005265135.1
Gene
UBE2E1
Gene Name
ubiquitin conjugating enzyme E2 E1
There are no transcripts associated with this gene.

View Full Product Details